Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercuric Chloride, 0.200 N (N/5), Ricca Chemical
SDP

Catalog No. 465516 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

Chemical Identifiers

CAS 7487-94-7, 7732-18-5
Molecular Formula Cl2Hg
Molecular Weight (g/mol) 271.49
MDL Number MFCD00011041
InChI Key LWJROJCJINYWOX-UHFFFAOYSA-L
Synonym mercuric chloride, mercury ii chloride, sublimate, mercury bichloride, mercury dichloride, corrosive sublimate, mercuric bichloride, mercury perchloride, sublimat, hgcl2
PubChem CID 24085
ChEBI CHEBI:31823
IUPAC Name mercury(2+) dichloride
SMILES [Cl-].[Cl-].[Hg++]

Specifications

Color Colorless
CAS Min % 95.0, 2.59
CAS Max % 99.0, 2.7
Grade Laboratory
Packaging Natural Poly Bottle
pH 3.2
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 0.200Normal
Chemical Name or Material Mercuric Chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.