Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercuric Chloride, ACS Reagent Mercury(II) Chloride, Reagents

Catalog No. 01337896
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Catalog No. Quantity
01-337-896 500 g
01-337-898 100 g
01-337-897 25 g
3 options

Catalog No. 01-337-896

Supplier: Reagents Holdings Llc C1245500500C2

Only null left
Add to Cart
Add to Cart

Mercuric Chloride, ACS Reagent Mercury(II) Chloride, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7487-94-7
Molecular Formula Cl2Hg
Molecular Weight (g/mol) 271.49
InChI Key LWJROJCJINYWOX-UHFFFAOYSA-L
Synonym Mercury(II) Chloride
IUPAC Name mercury(2+) dichloride
SMILES [Cl-].[Cl-].[Hg++]

Specifications

Boiling Point 302°C
Melting Point 276°C to 277 °C
Color Colorless/White
pH 3.2 to 4.7
Physical Form Solid
Quantity 500 g
Grade ACS
Packaging Glass Bottle
Specific Gravity 5.44
Chemical Name or Material Mercuric Chloride, ACS Reagent Mercury(II) Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.