Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercuric Chloride, Saturated, Certified, LabChem™
GSA_VA

Catalog No. lc166201
Click to view available options
Quantity:
500 mL
Packaging:
Poly Bottle

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 7487-94-7, 7732-18-5
Molecular Formula Cl2Hg
Molecular Weight (g/mol) 271.49
MDL Number MFCD00011041
InChI Key LWJROJCJINYWOX-UHFFFAOYSA-L
Synonym mercuric chloride, mercury ii chloride, sublimate, mercury bichloride, mercury dichloride, corrosive sublimate, mercuric bichloride, mercury perchloride, sublimat, hgcl2
PubChem CID 24085
ChEBI CHEBI:31823
IUPAC Name mercury(2+) dichloride
SMILES [Cl-].[Cl-].[Hg++]

Specifications

Color Colorless
CAS Max % 93.5, 6.5
Physical Form Liquid
Quantity 500 mL
Linear Formula HgCl2
UN Number UN2024
Solubility Information Soluble in water
Formula Weight 271.5
Grade Certified
Identification Passes Test
Packaging Poly Bottle
Specific Gravity >1.040
Decomposition Information Hydrogen chloride; Mercury
Chemical Name or Material Mercuric Chloride, Saturated
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Fatal if swallowed.
Toxic in contact with skin.
Suspected of causing cancer.
Suspected of damaging fertility or the unborn child.
May cause damage to organs (nervous system) through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not breathe mist.
Do not eat, drink or smoke when using this product.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse mouth.
Immediately call a poison center/doctor.
If on skin: Wash with plenty of soap and water.
Call a poison center/doctor if you feel unwell.
Remove/Take off immediately all contaminated clothing and wash it before reuse.
Collect spillage.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.