Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercuric Nitrate, 0.0100 N (N/100), Ricca Chemical
SDP

Catalog No. R470400020F Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
4 L
20 L
Packaging:
Amber Glass Bottle
Cubitainer
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R470400020F 20 L Cubitainer
R47040004C 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R470400020F Supplier Ricca Chemical Company Supplier No. R470400020F
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7732-18-5 , 7697-37-2 , 7783-34-8
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Color Colorless
Physical Form Liquid
Quantity 20 L
Assay Percent Range 0.01000 ± 0.00001 N at 20°C
Solubility Information Solubility in Water: Infinite
Grade Laboratory
Odor Odorless
Packaging Cubitainer
Chemical Name or Material Mercuric Nitrate, 0.0100 N (N/100)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.