Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercuric Nitrate, 0.0171 N, 0.00855 M, 1 mL = 1 mg NaCl, Ricca Chemical
SDP

Catalog No. R471000020F Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Amber Glass Bottle
Cubitainer
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R471000020F 20 L Cubitainer
4710-16 500 mL Amber Glass Bottle
4710-32 1 L Amber Glass Bottle
4710-1 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Catalog No. R471000020F Supplier Ricca Chemical Company Supplier No. R471000020F
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

Chemical Identifiers

CAS 7783-34-8 , 7697-37-2 , 7732-18-5
Molecular Formula H2HgN2O7
Molecular Weight (g/mol) 342.62
MDL Number MFCD00149736
InChI Key KVICROHOONHSRH-UHFFFAOYSA-N
Synonym mercuric nitrate monohydrate, unii-yj09edi5uy, mercury ii nitrate hydrate, yj09edi5uy, mercury 2+ ion hydrate dinitrate, nitric acid, mercury 2+ salt, monohydrate, mercury nitrate monohydrate, 324.60 anhydrous, mercury 2+ dinitrate hydrate, hgn2o6.xh2o x=1-2
PubChem CID 3084029
IUPAC Name mercury(2+) hydrate dinitrate
SMILES O.[Hg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Color Colorless
Equivalence 1 mL = 1 mg NaCl
Grade Laboratory
Packaging Cubitainer
Quantity 20 L
Physical Form Liquid
Concentration or Composition Notes 0.0171Normal,0.00855Molar
Chemical Name or Material Mercuric Nitrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.