Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercuric Sulfate Solution, APHA, Ricca Chemical
SDP

Catalog No. 478016 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
4780-16 500 mL Natural Poly Bottle
4780-32 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Catalog No. 4780-16 Supplier Ricca Chemical Company Supplier No. 478016
Add to Cart
Add to Cart
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

80 g/L Red Mercuric Oxide in 6 Normal Sulfuric Acid

Chemical Identifiers

CAS 7732-18-5 , 7664-93-9 , 21908-53-2

Specifications

Color Colorless
CAS Min % 70.0, 21.59, 6.21
CAS Max % 73.0, 22.47, 6.46
Grade Laboratory
Packaging Natural Poly Bottle
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 80g/L Red Mercuric Oxide in 6Normal Sulfuric Acid
Chemical Name or Material Mercuric Sulfate Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.