Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercuric Thiocyanate, 0.42% (w/v) in Methanol, Stock Solution for Chloride, Certified, LabChem™

Catalog No. lc167201
Click to view available options
Quantity:
500 mL
1 L

Chemical Identifiers

CAS 592-85-8, 67-56-1
Molecular Formula C4HgK2N4S4
Molecular Weight (g/mol) 511.10
InChI Key HWXOCCNUUYRBKP-UHFFFAOYSA-J
IUPAC Name mercury(2+) dipotassium tetrakis(cyanosulfanide)
SMILES [K+].[K+].[Hg++].[S-]C#N.[S-]C#N.[S-]C#N.[S-]C#N

Specifications

Color Colorless
CAS Max % 99.58, 0.42
Linear Formula Hg(CHNS)2
UN Number UN1993
Solubility Information Soluble in water
Formula Weight 316.75
Grade Certified
Identification Passes Test
Packaging Amber Glass
Suitability Passes Test
Decomposition Information Hydrogen cyanide; Carbon monoxide; Carbon dioxide; Mercury
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Mercuric Thiocyanate, 0.42% (w/v) in Methanol, Stock Solution for Chloride
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Toxic if swallowed or in contact with skin.
Fatal if inhaled.
Causes damage to organs (kidneys, liver, nervous system).
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, ventilating, lighting equipment.
Use only non-sparking tool.
Take precautionary measures against static discharge.
Do not breathe mist, vapors, spray.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, protective clothing, eye protection, face protection.
Wear respiratory protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If swallowed: Rinse mouth.
Iimmediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
In case of fire: Use alcohol resistant foam, extinguishing powder, dry sand to extinguish.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.