Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercurous Nitrate, Dihydrate, Crystal, Reagent, 97%, Spectrum™ Chemical
SDP

Catalog No. 18604322 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
500 g
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-322 12 kg Poly Pail
18-604-319 25 g Amber Glass Bottle
18-604-321 500 g Amber Glass Bottle
18-604-323 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-604-322 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M120812KG
Add to Cart
Add to Cart
Request a Quote

M1208, 14836-60-3, Hg2(NO3)2·2H2O

Spectrum™ Chemical Mercurous Nitrate, Dihydrate, Crystal, Reagent is also known as mercury nitrate, and is used in the preparation of other mercury compounds. This chemical compound, like all mercury compounds, is highly toxic. It has a white crystalline appearance, is noncombustible but will accelerate combustion due to its oxidizing properties. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.

Chemical Identifiers

CAS 14836-60-3
Molecular Formula H5HgNO5
Molecular Weight (g/mol) 299.63
InChI Key GJKBHHZLGNEESF-UHFFFAOYSA-N
IUPAC Name mercury(1+) nitric acid dihydrate
SMILES O.O.[Hg+].O[N+]([O-])=O

Specifications

CAS Max % 100%
Quantity 12 kg
UN Number UN1627
Percent Purity 97%
Grade Reagent
Packaging Poly Pail
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.