Learn More
Mercury(I) nitrate dihydrate, 97%
Description
Mercury(I) nitrate dihydrate is used in the preparation of other mercury(I) compounds. It acts as a reducing agent. It is involved in the preparation of mercury(II) nitrate by treating with dilute nitric acid as well as to get mercury on exposure to light.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Mercury(I) nitrate dihydrate is used in the preparation of other mercury(I) compounds. It acts as a reducing agent. It is involved in the preparation of mercury(II) nitrate by treating with dilute nitric acid as well as to get mercury on exposure to light.
Solubility
Slightly soluble in water.
Notes
Light sensitive. Incompatible with alcohols, sulfur and strong reducing agents.
Specifications
Specifications
| Melting Point | 70°C |
| Quantity | 50 g |
| Linear Formula | Hg2(NO3)2·2H2O |
| UN Number | UN1627 |
| Merck Index | 14,5896 |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 561.22 (525.19 Anhydrous) |
| Percent Purity | 98% |
| Odor | Odorless |
| Sensitivity | Light sensitive |
| Show More |
Safety and Handling
| Hazard Category | H300+H310+H330-H373 |
| Hazard Statement | GHS H Statement H300-H310-H330-H373 Fatal if swallowed. Fatal in contact with skin. Fatal if inhaled. May cause damage to organs through prolonged or repeated exposure. |
| Precautionary Statement | P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P314-P330-P361-P363-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: MERCUROUS NITRATE |
| EINECSNumber | 233-886-4 |
| RTECSNumber | OW8000000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.