Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercury(I) nitrate dihydrate, 98%, for analysis

Catalog No. p-44685
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Plastic bottle
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

CAS: 7782-86-7 | Hg2N2O6·2H2O | 561.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-86-7
Molecular Formula H4Hg2N2O8
Molecular Weight (g/mol) 561.22
MDL Number MFCD00150812
InChI Key LSABZDVKJBWCBE-UHFFFAOYSA-N
IUPAC Name dimercury(1+) dihydrate dinitrate
SMILES O.O.[Hg+].[Hg+].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 70.0°C
Color White to Yellow
Physical Form Crystalline Powder
Quantity 50 g
Assay Percent Range 97.5% min
Linear Formula Hg2(NO3)2·2H2O
Fieser 06,360
Merck Index 13, 5923
Solubility Information Solubility in water: 20g/L (20°C).
Formula Weight 561.22
Percent Purity 98%
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Mercury(I) nitrate dihydrate, p.a.
Show More Show Less
missing translation for 'healthHazard1' GHS Signal Word: Danger
missing translation for 'healthHazard2' GHS H Statement
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Very toxic to aquatic life with long lasting effects.
May cause damage to organs through prolonged or repeated exposure.
missing translation for 'healthHazard3' GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Gently wash w

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.