Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercury(I) nitrate dihydrate, ACS reagent, 9th edition

Catalog No. ac447180500
Click to view available options
Quantity:
50 g
250 g
Packaging:
Plastic bottle

CAS: 7782-86-7 | Hg2N2O6·2H2O | 561.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-86-7
Molecular Formula H4Hg2N2O8
Molecular Weight (g/mol) 561.22
InChI Key LSABZDVKJBWCBE-UHFFFAOYSA-N
IUPAC Name dimercury(1+) dihydrate dinitrate
SMILES O.O.[Hg+].[Hg+].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 70.0°C
Quantity 50 g
Linear Formula Hg2(NO3)2·2H2O
Solubility Information Solubility in water: 20g/L (20°C)
Formula Weight 561.22
Grade ACS Reagent
Packaging Plastic bottle
Chemical Name or Material Mercury(I) nitrate dihydrate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Very toxic to aquatic life with long lasting effects.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Gently wash w

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.