Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercury(II) nitrate monohydrate, 98+%

Catalog No. p-44748
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Plastic bottle
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

CAS: 7783-34-8 | H2HgN2O7 | 342.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-34-8
Molecular Formula H2HgN2O7
Molecular Weight (g/mol) 342.62
MDL Number MFCD00149736
InChI Key KVICROHOONHSRH-UHFFFAOYSA-N
Synonym mercuric nitrate monohydrate, unii-yj09edi5uy, mercury ii nitrate hydrate, yj09edi5uy, mercury 2+ ion hydrate dinitrate, nitric acid, mercury 2+ salt, monohydrate, mercury nitrate monohydrate, 324.60 anhydrous, mercury 2+ dinitrate hydrate, hgn2o6.xh2o x=1-2
PubChem CID 3084029
IUPAC Name mercury(2+) hydrate dinitrate
SMILES O.[Hg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 77.0°C to 79.0°C
Color White
Physical Form Adhering Crystalline Powder
Quantity 50 g
Assay Percent Range 98% min. (Complexometry)
Linear Formula Hg(NO3)2·H2O
Fieser 03,197; 10,254; 11,317
Merck Index 14, 5880
Solubility Information Solubility in water: slightly soluble
Formula Weight 342.6
Percent Purity ≥98%
Packaging Plastic bottle
Specific Gravity 4.3
Density 4.3000g/mL
Chemical Name or Material Mercury(II) nitrate monohydrate, p.a.
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause damage to organs through prolonged or repeated exposure.
Fatal if swallowed.
Very toxic to aquatic life with long lasting effects.
Fatal in contact with skin.
Fatal if inhaled.
Precautionary Statement GHS P Statement
Rinse mouth.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Re

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.