Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mercury(II) nitrate monohydrate, 98+%
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 77.0°C to 79.0°C |
Color | White |
Physical Form | Adhering Crystalline Powder |
Quantity | 250 g |
Assay Percent Range | 98% min. (Complexometry) |
Linear Formula | Hg(NO3)2·H2O |
Fieser | 03,197; 10,254; 11,317 |
Merck Index | 14, 5880 |
Solubility Information | Solubility in water: slightly soluble |
Formula Weight | 342.6 |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement May cause damage to organs through prolonged or repeated exposure. Fatal if swallowed. Very toxic to aquatic life with long lasting effects. Fatal in contact with skin. Fatal if inhaled. |
Precautionary Statement | GHS P Statement Rinse mouth. Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Gently wash with plenty of soap and water. Immediately call a POISON CENTER or doctor/physician. IF INHALED: Re |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction