Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercury(II) oxide, red, ACS, 99.0% min

Catalog No. AA1227736
Click to view available options
Quantity:
100 g
500 g

CAS: 21908-53-2 | HgO | 216.59 g/mol

It is used in marine and porcelain paints, dry batteries, chemical reagents, and topical antiseptics. It acts as a significant part in the production of mercury. It is utilitarian in the deduction of a mercury based superconductor. It is likewise employed, as a photo-oxidant for the photochemical oxidation of primary aromatic amines to the corresponding azoaromatic compounds. It is also an essential material for the preparation of cathodes in mercury batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21908-53-2
Molecular Formula HgO
Molecular Weight (g/mol) 216.59
MDL Number MFCD00011045
InChI Key UKWHYYKOEPRTIC-UHFFFAOYSA-N
Synonym mercuric oxide, mercury ii oxide, mercury oxide, mercuric oxide, red, red mercuric oxide, mercuric oxide, yellow, santar, red precipitate, mercury monoxide
IUPAC Name oxomercury
SMILES O=[Hg]

Specifications

Melting Point 500°C (decomposition)
Color Orange-Red
Physical Form Powder
Quantity 500 g
Assay Percent Range 99.0% min.
Solubility Information Partially soluble in water,insoluble in alcohol,ether,acetone,ammonia.
Formula Weight 216.59
Grade ACS
Odor Odorless
Appearance Dark orange powder
Density 11.14
Chemical Name or Material Mercury(II) oxide, red
Show More Show Less
Hazard Category H300+H310+H330-H373
Hazard Statement Keep away from food, drink and animal feeding stuffs. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.
Precautionary Statement P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P314-P330-P361-P363-P501c
DOTInformation DOT Class: 6.1, Packaging Group: II
EINECSNumber 244-654-7
RTECSNumber OW8750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.