Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercury(II) oxide, yellow, ACS, 99.0% min

Catalog No. p-4824816
Click to view available options
Quantity:
25 g
100 g

CAS: 21908-53-2 | HgO | 216.59 g/mol

Mercury(II) oxide production of mercury. It is also used as a material for cathodes for mercury batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21908-53-2
Molecular Formula HgO
Molecular Weight (g/mol) 216.59
MDL Number MFCD00011045
InChI Key UKWHYYKOEPRTIC-UHFFFAOYSA-N
Synonym mercuric oxide, mercury ii oxide, mercury oxide, mercuric oxide, red, red mercuric oxide, mercuric oxide, yellow, santar, red precipitate, mercury monoxide
IUPAC Name oxomercury
SMILES O=[Hg]

Specifications

Melting Point 500°C (decomposition)
Color Orange
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.0% min.
Solubility Information Insoluble in water. Insoluble in alcohol,ether,acetone,ammonia.
Formula Weight 216.59
Grade ACS
Appearance Dark orange
Density 11.14
Chemical Name or Material Mercury(II) oxide
Show More Show Less
Hazard Category H300+H310+H330-H373
Precautionary Statement P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P314-P330-P361-P363-P501c
DOTInformation DOT Class: 6.1; Packing Group: II
EINECSNumber 244-654-7
RTECSNumber OW8750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.