Learn More
Mercury(II) potassium iodide, Thermo Scientific Chemicals
Description
Mercury(II) potassium iodide is used as a pharmaceutical intermediate. It is also used as reagents, chemical research and organic intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Yellow |
| Physical Form | Crystalline Aggregates |
| Quantity | 250 g |
| Linear Formula | K2HgI4 |
| UN Number | UN2025 |
| Merck Index | 14,7688 |
| Solubility Information | Soluble in water. |
| Formula Weight | 786.4 |
| Sensitivity | Hygroscopic |
| Density | 4.3 g/mL |
| Show More |
Safety and Handling
| Hazard Category | H300+H310+H330-H373 |
| Hazard Statement | GHS H Statement H300-H310-H330-H373 Fatal if swallowed. Fatal in contact with skin. Fatal if inhaled. May cause damage to organs through prolonged or repeated exposure. |
| Precautionary Statement | P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P314-P330-P361-P363-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: MERCURY COMPOUNDS, SOLID, N.O.S. |
| EINECSNumber | 231-990-4 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.