Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercury(II) sulfide, Thermo Scientific Chemicals

Catalog No. p-4412848
Click to view available options
Quantity:
100 g

Mercury(II) sulfide, black is used chiefly as a pigment in paints, rubber and plastics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1344-48-5
Molecular Formula HgS
Molecular Weight (g/mol) 232.65
MDL Number MFCD00011046
InChI Key LGZXYFMMLRYXLK-UHFFFAOYSA-N
Synonym mercuric sulfide, mercury ii sulfide, vermilion, mercuric sulfide red, almaden, cinnabar, ethiops mineral, mercury sulfide, red cinnabar, mercury sulphide
PubChem CID 62402
SMILES [S--].[Hg++]

Specifications

Melting Point 583.5° (sublimation)
Color Black
Physical Form Powder
Quantity 100 g
Solubility Information Insoluble in water.
IUPAC Name mercury(2+) sulfanediide
Formula Weight 232.65
Odor Odorless
Appearance Black powder
Density 7.70 g/cm3
Chemical Name or Material Mercury(II) sulfide
Show More Show Less
Hazard Category H300+H310+H330-H373
Precautionary Statement P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P314-P330-P361-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 215-696-3
RTECSNumber OX0720000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.