Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Mercury(II) sulfide, Red, Alfa Aesar™

Click to view available options
Quantity:
100g
500g
1344-48-5
HgS
232.652
MFCD00011046
QXKXDIKCIPXUPL-UHFFFAOYSA-N
mercuric sulfide,mercury ii sulfide,vermilion,mercuric sulfide red,almaden,cinnabar,ethiops mineral,mercury sulfide,red cinnabar,mercury sulphide
62402
sulfanylidenemercury
S=[Hg]
1344-48-5
HgS
232.652
MFCD00011046
QXKXDIKCIPXUPL-UHFFFAOYSA-N
mercuric sulfide,mercury ii sulfide,vermilion,mercuric sulfide red,almaden,cinnabar,ethiops mineral,mercury sulfide,red cinnabar,mercury sulphide
62402
sulfanylidenemercury
S=[Hg]
Appearance Red powder
Solubility Information Insoluble in water.
Formula Weight 232.65
Physical Form Powder
Density 7.70 g/cm3
Odor Odorless
Color Red
Melting Point 583.5° (sublimation)
Quantity 100g
Chemical Name or Material Mercury(II) sulfide

P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P314-P330-P361-P363-P501c

H300+H310+H330-H373

DOTInformation : Hazard Class: 6.1; Packaging Group: II

EINECSNumber : 215-696-3

RTECSNumber : OX0720000

TSCA : Yes

Recommended Storage : Ambient temperatures