Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

m-Cresol Purple, sodium salt, pure, water soluble

Catalog No. AC199250050
Click to view available options
Quantity:
5 g
10 g
Packaging:
Glass bottle

CAS: 62625-31-4 | C21H17NaO5S

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62625-31-4
Molecular Formula C21H17NaO5S
MDL Number MFCD00010177
Synonym m-Cresol Purple, sodium salt

Specifications

Solubility Information Solubility in water: soluble.
Infrared Spectrum Authentic
Formula Weight 404.4
Grade Pure
Packaging Glass bottle
Visual Transition Interval from pH 7.4 (yellow) to pH 9.0 (purple)
Color Black to Brown/Orange
Quantity 5 g
Physical Form Powder
Chemical Name or Material m-Cresol Purple, sodium salt
EINECSNumber 263-656-9
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.