Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mercury Monoclonal Antibody (1H7)
GREENER_CHOICE

Catalog No. PIMA549688
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549688 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549688 Supplier Invitrogen™ Supplier No. MA549688
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Antibody tested in indirect ELISA.

Mercury is a naturally occurring element that can be found in air, water, and soil. It is a toxic substance that can cause serious health problems, including damage to the nervous system, kidneys, and lungs. Mercury can enter the environment through natural processes, such as volcanic activity and erosion, as well as through human activities, such as mining and industrial processes. It can accumulate in the food chain, particularly in fish and seafood, which can pose a risk to human health.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mercury
Applications ELISA, Immunohistochemistry, Lateral Flow, SDS-Page
Classification Monoclonal
Clone 1H7
Concentration 3.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Alias Hg
Host Species Mouse
Immunogen Conjugate of Hg hapten and KLH.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.