Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mercury(II) sulfate, Honeywell Fluka™
Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
100 g
250 g
3 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Quantity | 100 g |
| Linear Formula | HgSO4 |
| UN Number | UN1645 |
| Formula Weight | 296.65g/mol |
| Percent Purity | ≥99% (precipitation titration) |
| Chemical Name or Material | Mercury(II) sulfate |
Safety and Handling
| Hazard Statement | P260-P262-P280-P302 + P352 + P310-P304 + P340 + P310 |
| RTECSNumber | OX0500000 |
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction