Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Mercury(II) Thiocyanate

Catalog No. M4128100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Click to view available options
Quantity:
5 g
100 g

Catalog No. M4128100G

Supplier: Sigma Aldrich M4128100G

Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Mercury(II) Thiocyanate

Specifications

CAS 89-25-8
Melting Point 126°C to 128°C
Boiling Point 287°C (265 mmHg, lit.)
Molecular Formula C10H10N2O
Linear Formula C10H10N2O
MDL Number MFCD00003138
Quantity 5 g
Synonym Mercuric isothiocyanate; Mercuric sulfocyanide; Mercuric thiocyanate; Mercury di(thiocyanate); Thiocyanic acid, mercury(II) salt
Molecular Weight (g/mol) 174.2 g/mol
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.