Learn More
Meridian Bioscience™ Diagnostics Premier™ Adenoclone

Qualitative ELISA for the detection of human adenoviruses
$1088.00 - $1203.00
Specifications
Certifications/Compliance | CE Marked, FDA |
---|---|
Content And Storage | 2°C to 8°C |
CPT Code | 87301 |
Type | Adenovirus Test Kit |
Includes | 48 Microwells, 50mL sample diluent/negative control, 60mL Enzyme conjugate, 2mL positive control, 6mL substrates and stop solution, microwell holder, 48 sample transfer pipettes |
Catalog Number | Mfr. No. | Sample Type | Sensitivity | Specificity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Sample Type | Sensitivity | Specificity | Price | Quantity & Availability | |||
14-910-181
![]() |
Meridian Bioscience™
696006 |
Cell Culture, Stool | 98% | 0.98 |
Each for $1,203.00
|
|
|||
14-910-180
![]() |
Meridian Bioscience™
696007 |
Stool | 99% | 1 |
Each for $1,088.00
|
|
|||
Description
- Sensitivity: 98%
- Specificity: 98%
- Enzyme immunoassay that utilizes monoclonal antibodies
- Allows qualitative detection of human adenoviruses direct from stool specimens
- Results Interpretation: Spectrophotometer/Visual
- For in vitro use only
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
CE Marked, FDA | |
87301 | |
48 Microwells, 50mL sample diluent/negative control, 60mL Enzyme conjugate, 2mL positive control, 6mL substrates and stop solution, microwell holder, 48 sample transfer pipettes | |
75 min. |
2°C to 8°C | |
Adenovirus Test Kit | |
48 Tests |
Safety and Handling
ShelfLife : 18 Months
Your input is important to us. Please complete this form to provide feedback related to the content on this product.