Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meridian Bioscience™ Illumigene Mycoplasma Direct DNA Amplification Assay

Catalog No. 23315124 Shop All Meridian Bioscience Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-315-124 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-315-124 Supplier Meridian Bioscience™ Supplier No. 280250

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Meridian Bioscience™ Illumigene Mycoplasma Direct DNA Amplification Assay is a qualitative in vitro diagnostic test for detection of Mycoplasma pneumoniae, DNA in human throat swabs obtained from patients suspected of having M. pneumoniae infection. Uses loop-mediated isothermal DNA amplification (LAMP) technology to target segment of the M. pneumoniae genome.

illumigene kits were rebranded as Alethia. Product is identical only the name is different. No validation of Alethia is needed if currently using illumigene version. If discontinued illumigene kits are ordered the Alethia version will ship. New Number is  23029012

  • Results in less than 1 hour
  • Detects Mycoplasma pneumoniae on day 1 of symptoms
  • Simple sample preparation from throat swabs
  • High sensitivity and specificity
  • 0.0% Invalid Rate
  • Quicker initiation of appropriate patient care
  • Expanded molecular menu
  • No capital expense or service contract required
  • Small footprint, easily adaptable to any laboratory

Specifications

Certifications/Compliance CE Marked, FDA Cleared
Content And Storage 2°C to 27°C
Description FDA Cleared
CE Marker Yes
Clia Complexity Moderate
Sample Type Throat Swab
Sensitivity 96%
Quantity 50 Tests
Specificity 0.977
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.