Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meridian Bioscience™ ImmunoCard STAT!™ Campy
GSA_VA

Catalog No. 23315090 Shop All Meridian Bioscience Products
Change view
Click to view available options
Quantity:
30 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-315-090 30 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-315-090 Supplier Meridian Bioscience™ Supplier No. 751530
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

For the detection of Campylobacter

  • Results in 20 minutes
  • Allows for triage of patient for earlier therapy
  • 98.1% Sensitivity, 95.9% specificity
  • Greater sample flexibility, culture detects only viable organisms
  • Antibiotics in Campylobacter culture media can suppress growth

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

CPT Code 87899
Detectable Analytes Campylobacter Antigen
Format Card
Sample Type Human Stool
Type ImmunoCard Stat™ Campy Test
Quantity 30 Tests
Test Time 20 min.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.