Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meridian Bioscience™ ImmunoCard™ STAT! Rotavirus Test Kit
GSA_VA

Catalog No. 14910172 Shop All Meridian Bioscience Products
Change view
Click to view available options
Quantity:
30 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-910-172 30 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 14-910-172 Supplier Meridian Bioscience™ Supplier No. 750030
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

30 individually wrapped test devices, positive control, sample diluent, and 30 transfer pipets

No centrifugation required; simply dilute specimen and add to test device

  • CLIA Complexity: Moderate
  • Rapid membrane test for the qualitative detection of rotavirus antigen in human stool
  • Results in less than 10 minutes
  • Sensitivity: 93.1%; specificity: 95.8%

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Certifications/Compliance CE Marked
Content And Storage 2°C to 8°C
CPT Code 87425
Clia Complexity Moderate
Detectable Analytes Rotavirus Antigen
Sensitivity 93.1%
Type ImmunoCard Stat™ Rotavirus Test Kit
Includes 30 Individually wrapped test devices, positive control, sample diluent and 30 transfer pipets
Quantity 30 Tests
Specificity 0.958
Test Time 10 min.
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.