Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MERIT40/HSPC142 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00
Specifications
| Antigen | MERIT40/HSPC142 |
|---|---|
| Dilution | Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
MERIT40/HSPC142 Polyclonal antibody specifically detects MERIT40/HSPC142 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| MERIT40/HSPC142 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| DNA replication Transcription Translation and Splicing | |
| PBS (pH 7.2), 40% Glycerol | |
| 29086 | |
| IgG | |
| Immunogen affinity purified |
| Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| BRISC and BRCA1 A complex member 1, C10orf62, C19orf62, chromosome 19 open reading frame 62, FLJ20571, HSPC142, mediator of Rap80 interactions and targeting 40 kDa, Mediator of RAP80 interactions and targeting subunit of 40 kDa, MERIT40BRCA1-A complex subunit MERIT40, NBA1BRISC and BRCA1 A complex member, new component of the BRCA1 A complex, New component of the BRCA1-A complex, new component of the BRCAA1 A complex | |
| This antibody was developed against a recombinant protein corresponding to amino acids: SLPKLESFNGSKTNALNVSQKMIEMFVRTKHKIDKSHEFALVVVNDDTAWLSGLTSDPRELCSCLYDLETASCSTFNLEGL | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title