Learn More
MERIT40 Polyclonal Antibody, Bethyl Laboratories
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat, Bovine, Dog, Horse, Pig, Panda, Orangutan, Monkey, Gorilla, Chimpanzee, Chinese hamst
Specifications
Specifications
| Antigen | MERIT40 |
| Applications | Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.1% BSA and 0.09% sodium azide |
| Gene | BABAM1 |
| Gene Accession No. | Q9NWV8 |
| Gene Alias | C19orf62, MERIT40, NBA1, BRCA1-A complex subunit MERIT40, BRISC and BRCA1-A complex member 1, mediator of RAP80 interactions and targeting subunit of 40 kDa, mediator of Rap80 interactions and targeting 40 kDa, new component of the BRCA1 A complex, new component of the BRCA1-A complex |
| Gene Symbols | BABAM1 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.