Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Merlin (NF2), Mouse, Unlabeled, Clone: 1C4,

Catalog No. MABN1786 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABN1786 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN1786 Supplier MilliporeSigma™ Supplier No. MABN1786
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Merlin (UniProt P35240; also known as Moesin-ezrin-radixin-like protein, Neurofibromin-2, Schwannomerlin, Schwannomin) is encoded by the NF2 (also known as SCH) gene (Gene ID 4771) in human. Originally identified as a tumor suppressor gene in the familial Neurofibromatosis type 2 (NF2) cancer patients, NF2 encodes the FERM (Ezrin, Radixin, Moesin) domain-containing merlin that plays a negative regulating role against cell growth and proliferation by functioning as an upstream activator of the Hippo pathway. Merlin directly activates Mst1/2 and recruits Lats1/2 to the plasma membrane for phosphorylation by Mst1/2. The phosphorylated Lats1/2 in turn inhibits YAP/TAZ transcription activity by phosphorylating YAP/TAZ and causes its cytoplasmic sequestration. Merlin also translocates to the nucleus, where it inhibits E3 ubiquitin ligase CRL4(DCAF1), thereby preventing it from ubiquitinating and downregulating Lats1/2. In its inactive state, the C-terminal tail domain (CTD; a.a. 507-595) of merlin is bound intramolecularly to its FERM domain (a.a. 19-313). Angiomotin (AMOT) induces merlin activation by binding to an elongated helical fragment within merlin CTD and subsequently releases the auto-inhibited conformation of Merlin. Merlin CTD can also undergo phosphorylation on Ser518 by p21-activated kinase (PAK) or cyclic AMP-dependent protein kinase (PKA), resulting in a weakened AMOT binding and a stabilization of merlin in its auto-inhibited state.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Merlin (NF2)
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 1C4
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse IgG1 in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene NF2, SCH
Gene Accession No. P35240
Host Species Mouse
Immunogen Recombinant protein corresponding to the C-terminal half of human merlin (NF2) isoform II (UniProt spliced isoform 3; P35240-3).
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_057502
Test Specificity Clone 1C4 detected merlin (NF2), but not any of the three ERM proteins, ezrin, radixin, and moesin (Gonzalez-Agosti, C., et al. (1996). Oncogene. 13(6):1239-1247). Clone 1C4 targeted epitope is present in human merlin (NF2) spliced isoforms 1 to 6, but not 7, 9, or 10 as reported by UniProt (P35240). Reactivity toward human spliced isoform 8 is possible, but has not been confirmed.
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.