Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meropenem Trihydrate, 98-101%, PROMO, Thermo Scientific Chemicals

Catalog No. J66446MFPM
Click to view available options
Quantity:
50 mg

Applications

Meropenem trihydrate is an ultra-broad spectrum beta-lactam antibiotic active against both Gram-positive and Gram-negative bacteria.

Solubility

Soluble in aqueous solution at approximately 5 mg/mL

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119478-56-7
Molecular Formula C17H25N3O5S
Molecular Weight (g/mol) 383.46 g/mol
MDL Number MFCD08600005,MFCD00864966
InChI Key FSTGLKRHSQANLP-PQTSNVLCSA-N
PubChem CID 441130
ChEBI CHEBI:43968
IUPAC Name (4R,5S,6S)-3-{[(2S,5S)-5-(dimethylcarbamoyl)pyrrolidin-2-yl]sulfanyl}-6-[(1R)-1-hydroxyethyl]-4-methyl-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid
SMILES C[C@@H](O)[C@@H]1[C@H]2[C@@H](C)C(S[C@H]3CC[C@H](N3)C(=O)N(C)C)=C(N2C1=O)C(O)=O

Specifications

Type Meropenem Trihydrate
Physical Form Powder
Color White to Yellow
Quantity 50 mg
Flash Point >110°C (230°F)
Boiling Point 627°C
Merck Index 14,5900
Solubility Information Soluble in aqueous solution
Chemical Name or Material Meropenem trihydrate
RTECSNumber CL5446509
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.