Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MERS Coronavirus Envelope Protein Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5143455
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
Middle East Respiratory Syndrome (MERS) is a contagious viral respiratory infection, whose origin was identified as the MERS-related coronavirus (MERS-CoV) MERS-CoV is a single-stranded mRNA betacoronavirus The MERS-CoV genome encodes four key structural proteins (spike, envelope, membrane, and nucleocapsid) that make up the virion particle The MERS envelope protein (E) is the smallest of the structural proteins, and is abundantly expressed in the host cell during viral replication, despite only a fraction of synthesized E protein ultimately being incorporated into new viral particles.
Specifications
MERS Coronavirus Envelope Protein | |
Polyclonal | |
Unconjugated | |
Coronavirus; Envelope protein; MERS | |
Recombinant protein encompassing a sequence within the center region of MERS-CoV Envelope protein. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1.25 mg/mL | |
PBS with 20% glycerol and 0.025% Proclin 300; pH 7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
Virus | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction