Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MERS Coronavirus NP Monoclonal Antibody (10)
GREENER_CHOICE

Catalog No. PIMA529972
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA529972 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA529972 Supplier Invitrogen™ Supplier No. MA529972
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for The antibody reacts with MERS-CoV (NCoV/Novel coronavirus) Nucleocapsid protein (NP protein)..

Packages the positive strand viral genome RNA into a helical ribonucleocapsid (RNP and plays a fundamental role during virion assembly through its interactions with the viral genome and membrane protein M. Plays an important role in enhancing the efficiency of subgenomic viral RNA transcription as well as viral replication.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MERS Coronavirus NP
Applications ELISA, Western Blot
Classification Monoclonal
Clone 10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Alias Coronavirus; MERS; NP
Host Species Mouse
Immunogen Recombinant MERS-CoV (NCoV/Novel coronavirus) Nucleoprotein/NP protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.