Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MERS Coronavirus NP Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5112051
Description
Specificity: The antibody reacts with MERS-CoV (NCoV / Novel coronavirus) Nucleocapsid protein (NPprotein).
Packages the positive strand viral genome RNA into a helical ribonucleocapsid (RNP and plays a fundamental role during virion assembly through its interactions with the viral genome and membrane protein M. Plays an important role in enhancing the efficiency of subgenomic viral RNA transcription as well as viral replication.
Specifications
| MERS Coronavirus NP | |
| Polyclonal | |
| Unconjugated | |
| Coronavirus; MERS; NP | |
| A synthetic peptide corresponding to the center region of MERS-CoV (NCoV/Novel coronavirus) Nucleocapsid Protein (NP protein). | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with no preservative | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction