Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MERS-CoV Nucleocapsid Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB10729100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB10729100 100 μg
MAB10729SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB10729100 Supplier R&D Systems Supplier No. MAB10729100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Nucleocapsid Monoclonal antibody specifically detects Nucleocapsid in MERS-CoV samples. It is validated for Western Blot, Western Blot

Specifications

Antigen Nucleocapsid
Applications Western Blot
Classification Monoclonal
Clone 1038729
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Simple Western 25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. YP_007188586.1
Host Species Mouse
Immunogen Spodoptera frugiperda insect ovarian cell line Sf21-derived mouse MERS-CoV Nucleocapsid, Met1-Thr411, Accession # YP_007188586.1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species MERS-CoV
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.