Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MERS-CoV Spike Protein Antibody (12), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP326065AF750
Description
MERS-CoV Spike Protein Monoclonal antibody specifically detects MERS-CoV Spike Protein in Virus samples. It is validated for Western Blot, ELISASpecifications
MERS-CoV Spike Protein | |
Monoclonal | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Virus | |
Purified |
Western Blot, ELISA | |
12 | |
Western Blot, ELISA | |
MERS S glycoprotein, MERS S1, MERS S2, MERS Spike glycoprotein, MERS Spike Protein, MERS-CoV S glycoprotein, Middle East respiratory syndrome-related coronavirus Spike glycoprotein, Spike glycoprotein, spike protein, Spike RBD, surface glycoprotein | |
Hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant MERS-CoV Spike Protein (Accession#: AFS88936.1, Met1-Trp1297) | |
0.1 mL | |
Infections (Virus Bacteria and Parasites) | |
14254594 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction