Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MERS-CoV Spike Protein Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP305814B
Description
MERS-CoV/NCov Spike Protein Polyclonal specifically detects MERS-CoV/NCov Spike Protein in MERS-CoV samples. It is validated for Western Blot.Specifications
MERS-CoV/NCov Spike Protein | |
Polyclonal | |
Western Blot | |
coronavirus s1, coronavirus s2, coronavirus spike, cov spike, E2, Human coronavirus spike glycoprotein, MERS, MERS Peplomer protein, MERS S glycoprotein, MERS S1, MERS S2, MERS Spike glycoprotein, MERS Spike Protein, MERS-CoV E2, MERS-CoV Peplomer protein, MERSCoV S glycoprotein, MERS-CoV S glycoprotein, MERSCoV Spike glycoprotein, MERS-CoV Spike glycoprotein, MERSCOV Spike Protein, Middle East respiratory syndrome-related coronavirus Spike glycoprotein, ncov RBD, ncov s1, ncov s2, ncov spike, Peplomer proteinMERS-CoV Spike glycoprotein, RBD, S, S1, Spike glycoprotein, spike protein, Spike RBD, surface glycoprotein | |
Produced in rabbits immunized with purified, a synthetic peptide corresponding to the center region of the S1 subunit of MERS-CoV/NCov Spike Protein (S protein). | |
0.1 mL | |
Primary | |
MERS-CoV | |
Purified |
Western Blot | |
Biotin | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
14254594 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction