Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MERS-CoV Spike Protein Rabbit anti-MERS-CoV, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$274.50 - $469.50
Specifications
| Antigen | MERS-CoV Spike Protein |
|---|---|
| Dilution | Western Blot, ELISA 1:5000-1:10000 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
MERS-CoV Spike Protein Polyclonal antibody specifically detects MERS-CoV Spike Protein in MERS-CoV samples. It is validated for Western Blot, ELISASpecifications
| MERS-CoV Spike Protein | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Infections (Virus Bacteria and Parasites) | |
| 0.2 um filtered solution in PBS | |
| 14254594 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot, ELISA 1:5000-1:10000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| MERS-CoV | |
| MERS S glycoprotein, MERS S1, MERS S2, MERS Spike glycoprotein, MERS Spike Protein, MERS-CoV S glycoprotein, Middle East respiratory syndrome-related coronavirus Spike glycoprotein, Spike glycoprotein, spike protein, Spike RBD, surface glycoprotein | |
| Produced in rabbits immunized with purified recombinant MERS-CoV Spike Protein (Asp726-Pro1296, Accession: AFS88936.1) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title