Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MERS-CoV Spike RBD Alexa Fluor™ 700-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB107073N1
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB107073N1 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB107073N1 Supplier R&D Systems Supplier No. FAB107073N100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

SARS-CoV-2 Spike RBD Monoclonal specifically detects SARS-CoV-2 Spike RBD in MERS-CoV samples. It is validated for Flow Cytometry.

Specifications

Antigen SARS-CoV-2 Spike RBD
Applications Flow Cytometry
Classification Monoclonal
Clone 1038345
Conjugate Alexa Fluor 700
Dilution Flow Cytometry 0.25-1 ug/106 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. YP_007188579.1
Gene Alias 2019-nCoV Peplomer protein, 2019-nCoV S Protein, 2019-nCoV Spike, COVID-19 Spike, E2, Human coronavirus spike glycoprotein, Peplomer protein, S glycoprotein, S protein, SARS-CoV-2, SARS-COV-2 E2, SARS-COV-2 Peplomer protein, SARS-COV-2 S protein, SARS-COV-2 Spike glycoprotein, SARSCOV2 Spike protein, Severe Acute Respiratory Syndrome Coronavirus 2 Spike Protein, Spike, Spike glycoprotein, Spike S1, Spike S1 RBD, surface glycoprotein
Host Species Mouse
Immunogen Chinese Hamster Ovary cell line CHO-derived MERS-CoV Spike RBD, Glu367-Tyr606, Accession # YP_007188579.1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 43740568
Reconstitution Reconstitution default not known
Target Species MERS-CoV
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.