Learn More
Invitrogen™ MERTK Recombinant Rabbit Monoclonal Antibody (4I1P7)

Description
Immunogen sequence: MGPAPLPLLL GLFLPALWRR AITEAREEAK PYPLFPGPFP GSLQTDHTPL LSLPHASGYQ PALMFSPTQP GRPHTGNVAI PQVTSVESKP LPPLAFKHTV.
Specifications
Specifications
| Antigen | MERTK |
| Applications | ELISA, Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | 4I1P7 |
| Concentration | 0.75 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | Mertk |
| Gene Accession No. | Q12866 |
| Gene Alias | c-Eyk; c-mer; c-mer proto-oncogene tyrosine kinase; Eyk; MER; MER proto-oncogene, tyrosine kinase; MER receptor tyrosine kinase; MERTK; MGC133349; nmf12; Nyk; Proto-oncogene c-Mer; proto-oncogene tyrosine-protein kinase MER; rdy; receptor tyrosine kinase MerTK; Receptor tyrosine tinase gene probably the gene for Rdy; retinal dystrophy; RP38; sMER; sMERTK; soluble MER; soluble MERTK; STK kinase; Tyro 12; Tyro12; Tyrosine-protein kinase Mer |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.