Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MES, 0.2 M Buffer Solution, pH 6.0, PROMO, Thermo Scientific Chemicals

Catalog No. J60930APPM
Click to view available options
Quantity:
500 mL

Applications

MES is used as a running buffer for resolving very small proteins (ex: 13 kDa) on bis-tris gels. Also used in cation-exchange chromatography and as a biological buffer in plant cell cultures. Used in various protein characterization assays. MES is also useful as a non-coordinating buffer in chemistry involving metal ions, as many common buffers (e.g. phosphate and acetate) readily form coordination complexes. MES only weakly binds Ca, Mg, Mn, and it has negligible binding with Cu(II).

Solubility

Soluble in water (approx.1.3 M).

Notes

Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 145224-94-8
Molecular Formula C6H15NO5S
Molecular Weight (g/mol) 213.25 g/mol
MDL Number MFCD00283848
InChI Key MIIIXQJBDGSIKL-UHFFFAOYSA-N
PubChem CID 16218417
IUPAC Name 2-morpholin-4-ylethanesulfonic acid;hydrate
SMILES C1COCCN1CCS(=O)(=O)O.O

Specifications

Color Colorless
Concentration 0.2 M
pH 6.0
Physical Form Liquid
Quantity 500 mL
Solubility Information Soluble in water.
Chemical Name or Material MES buffer solution

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.