Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals MES, 0.5M buffer soln., pH 5.0

Catalog No. p-7042308
Click to view available options
Quantity:
100 mL
250 mL

CAS: 145224-94-8 | C6H15NO5S | 213.248 g/mol

MES, is often used as a running buffer for resolving very small proteins on bis-tris gels, also used as a biological buffer in plant cell cultures. It is also been used in various protein characterization assays. This sterile solution suitable for cell culture and molecular biology applications. It is also used as a buffering agent in biology and biochemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
MES, is often used as a running buffer for resolving very small proteins on bis-tris gels, also used as a biological buffer in plant cell cultures. It is also been used in various protein characterization assays. This sterile solution suitable for cell culture and molecular biology applications. It is also used as a buffering agent in biology and biochemistry.

Solubility
Soluble in water (0.5 M at 20°C).

Notes
Solutions are stable at 2-8°C for months. Sterilize by filtration through 0.2 mm filters. Autoclaving is not recommended for any sulfonic acid buffer. If buffers must be nuclease-free, treat the water first, then add the buffer after autoclaving. When MES solutions are autoclaved, they turn yellow (although pH does not change measurably). The identity of the yellow breakdown product is unknown.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 145224-94-8
Molecular Formula C6H15NO5S
Molecular Weight (g/mol) 213.248
MDL Number MFCD00283848
InChI Key MIIIXQJBDGSIKL-UHFFFAOYSA-N
PubChem CID 16218417
IUPAC Name 2-morpholin-4-ylethanesulfonic acid;hydrate
SMILES C1COCCN1CCS(=O)(=O)O.O

Specifications

Color Colorless
Concentration 0.5 M
pH 5.0
Physical Form Liquid
Quantity 100 mL
Solubility Information Soluble in water (0.5 M at 20°C).
Chemical Name or Material MES buffer soln.
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.