Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MES, 1.0 M Buffer Solution, pH 9.0, Thermo Scientific™

Catalog No. J61656AKPM
Click to view available options
Quantity:
250 mL

Applications

MES is used as a buffering agent in biology and biochemistry. It is utilized for resolving very small proteins on bis-tris gels. It is mainly used as a biological buffer in plant cell cultures. Buffer solutions are used as a means of keeping pH at a nearly constant value in a wide variety of chemical applications.

Solubility

Miscible with water.

Notes

Incompatible with strong oxidizing agents.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 145224-94-8
Molecular Formula C6H15NO5S
Molecular Weight (g/mol) 213.25 g/mol
MDL Number MFCD00283848
InChI Key MIIIXQJBDGSIKL-UHFFFAOYSA-N
PubChem CID 16218417
IUPAC Name 2-morpholin-4-ylethanesulfonic acid;hydrate
SMILES C1COCCN1CCS(=O)(=O)O.O

Specifications

Concentration 1.0 M
pH 9.0
Physical Form Liquid
Quantity 250 mL
Solubility Information Fully miscible.
Chemical Name or Material MES buffer solution

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.