Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MES Hydrate, 99+%, PROMO, Thermo Scientific Chemicals

Catalog No. H5647236PM
Click to view available options
Quantity:
500 g

Applications

MES hydrate is a zwitterionic N-substituted aminosulfonic acid, known as Good's buffer, is utilized for many biological applications. It is a useful resolving agent for very small proteins on bis-tris gels. It is mainly used as a biological buffer in plant cell cultures. Buffer solutions are used as a means of keeping pH at a nearly constant value in a wide variety of chemical applications.

Solubility

Soluble in water. Slightly soluble in organic solvents.

Notes

Hygroscopic. Incompatible with strong oxidizing agents.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1266615-59-1
Molecular Formula C6H13NO4S
Molecular Weight (g/mol) 195.23 g/mol
MDL Number MFCD00149409
InChI Key SXGZJKUKBWWHRA-UHFFFAOYSA-N
Synonym 2-(4-Morpholinyl)ethanesulfonic acid hydrate; 4-Morpholineethanesulfonic acid hydrate
IUPAC Name 2-(morpholin-4-yl)ethane-1-sulfonic acid
SMILES OS(=O)(=O)CCN1CCOCC1

Specifications

Melting Point Approx. 308°C (decomposition)
Quantity 500 g
Beilstein 141319
Merck Index 14,5902
Solubility Information Soluble in water. Slightly soluble in organic solvents.
Percent Purity ≥99%
Chemical Name or Material MES hydrate
EINECSNumber 224-632-3
RTECSNumber QE3479500
TSCA Yes

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.