Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MES-SDS Running Buffer (20X), PROMO, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 L
Description
Applications
MES-SDS running buffer (20X) is used for high resolution of proteins on neutral pH. It finds application in the separation of small to medium-sized proteins.
Solubility
Miscible with water.
Notes
This buffer contains: 1 M MES, 1 M Tris base, 2% SDS and 20mM EDTA at pH 7.3. Store at room temperature. Keep away from light. Incompatible with strong oxidizing agent and acids.

Specifications
Specifications
Concentration | 20X |
Physical Form | Liquid |
Quantity | 1 L |
RUO - Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction