Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-1,4-Dibromo-2-butene, 97%

Catalog No. AC406465000
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle

CAS: 821-06-7 | C4H6Br2 | 213.9 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 821-06-7
Molecular Formula C4H6Br2
Molecular Weight (g/mol) 213.9
MDL Number MFCD00000249
InChI Key RMXLHIUHKIVPAB-OWOJBTEDSA-N
Synonym 1,4-dibromo-2-butene, trans-1,4-dibromo-2-butene, e-1,4-dibromobut-2-ene, e-1,4-dibromo-2-butene, 1,4-dibromobut-2-ene, trans-1,4-dibromobut-2-ene, 2-butene, 1,4-dibromo, 2-butene, 1,4-dibromo-, 2e, 2e-1,4-dibromobut-2-ene, 1,4-dibromo-2-butylene
PubChem CID 641245
IUPAC Name (E)-1,4-dibromobut-2-ene
SMILES C(C=CCBr)Br

Specifications

Melting Point 48°C to 53°C
Color White to Beige
Boiling Point 205°C
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula BrCH2CHCHCH2Br
Quantity 500 g
Solubility Information Solubility in water: slightly soluble. Other solubilities: slightly soluble in chloroform,soluble in ethanol,acetone
Formula Weight 213.9
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material trans-1, 4-Dibromo-2-butene, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Toxic if inhaled.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 212-472-7
Recommended Storage Refrigerator +4°C

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.