Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MESDC1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584871
Description
Immunogen sequence: LLTQCLRDLA QHPDGGAKMS DHRERLRNSA CAVSEGCTLL SQALRERSSP RTLPPVNSNS VN.
Actin-binding protein which may have an oncogenic function and regulates cell proliferation, migration and invasion in cancer cells.
Specifications
MESDC1 | |
Polyclonal | |
Unconjugated | |
Tlnrd1 | |
AW061151; MESDC1; Mesoderm development candidate 1; mesoderm development candidate 1; talin rod domain containing 1; talin rod domain containing 1; talin rod domain-containing protein 1; TLNRD1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
59274 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunofluorescence | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H1K6 | |
Tlnrd1 | |
Recombinant protein corresponding to Human MESDC1. Recombinant protein control fragment (Product #RP-108587). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction