Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MESDC2 Monoclonal Antibody (S.539.9)
GREENER_CHOICE

Catalog No. PIMA514906
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA514906 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA514906 Supplier Invitrogen™ Supplier No. MA514906
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

It is not recommended to aliquot this antibody.

MESDC2 chaperone specifically assisting the folding of beta-propeller/EGF modules within the family of low-density lipoprotein receptors (LDLRs). Acts as a modulator of the Wnt pathway through chaperoning the coreceptors of the canonical Wnt pathway, LRP5 and LRP6, to the plasma membrane. Essential for specification of embryonic polarity and mesoderm induction. Plays an essential role in neuromuscular junction (NMJ) formation by promoting cell-surface expression of LRP4. May regulate phagocytosis of apoptotic retinal pigment epithelium (RPE) cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MESDC2
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone S.539.9
Concentration 35 μg/mL
Conjugate Unconjugated
Formulation 0.01M HEPES with 50% glycerol, 100μg/mL BSA, 0.15M NaCl and <0.02% sodium azide; pH 7.5
Gene Mesd
Gene Accession No. Q14696, Q5U2R7, Q9ERE7
Gene Alias 2210015O11Rik; AW537813; BOCA; KIAA0081; LDLR chaperone MESD; LRP chaperone MESD; MESD; Mesdc2; MESDM; mesoderm development candiate 2; mesoderm development candidate 2; Mesoderm development LRP chaperone MESD; Mesoderm development protein; mKIAA0081; msd; renal carcinoma antigen NY-REN-61; UNQ1911/PRO4369
Gene Symbols Mesd
Host Species Rabbit
Immunogen Recombinant human MESDC2 protein.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23184, 308796, 67943
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.