Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MESDC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5118057
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5118057 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5118057 Supplier Invitrogen™ Supplier No. PA5118057
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MESDC2 Polyclonal Antibody for Western Blot, IP

MESDC2 chaperone specifically assisting the folding of beta-propeller/EGF modules within the family of low-density lipoprotein receptors (LDLRs). Acts as a modulator of the Wnt pathway through chaperoning the coreceptors of the canonical Wnt pathway, LRP5 and LRP6, to the plasma membrane. Essential for specification of embryonic polarity and mesoderm induction. Plays an essential role in neuromuscular junction (NMJ) formation by promoting cell-surface expression of LRP4. May regulate phagocytosis of apoptotic retinal pigment epithelium (RPE) cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MESDC2
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene Mesd
Gene Accession No. Q14696
Gene Alias 2210015O11Rik; AW537813; BOCA; KIAA0081; LDLR chaperone MESD; LRP chaperone MESD; MESD; Mesdc2; MESDM; mesoderm development candiate 2; mesoderm development candidate 2; Mesoderm development LRP chaperone MESD; Mesoderm development protein; mKIAA0081; msd; renal carcinoma antigen NY-REN-61; UNQ1911/PRO4369
Gene Symbols Mesd
Host Species Rabbit
Immunogen E. coli-derived Human MESDC2 fragment.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23184
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.