Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mesitaldehyde, 97%

Catalog No. p-156204
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle

CAS: 487-68-3 | C10H12O | 148.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 487-68-3
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.21
MDL Number MFCD00003341
InChI Key HIKRJHFHGKZKRI-UHFFFAOYSA-N
Synonym mesitaldehyde, mesitylaldehyde, benzaldehyde, 2,4,6-trimethyl, 2-mesitylenecarboxaldehyde, mesitylenecarboxaldehyde, 2-formylmesitylene, 2,4,6-trimethyl-benzaldehyde, isodural, mesityl aldehyde, b-isodural
PubChem CID 10254
IUPAC Name 2,4,6-trimethylbenzaldehyde
SMILES CC1=CC(C)=C(C=O)C(C)=C1

Specifications

Melting Point 14.0°C
Color Colorless to Yellow
Density 1.0000g/mL
Boiling Point 237.0°C
Flash Point 105°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5520 to 1.5540
Linear Formula (CH3)3C6H2CHO
Quantity 10 g
Beilstein 07, 325
Specific Gravity 1
Formula Weight 148.2
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Mesitaldehyde, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 207-662-1
RTECSNumber CU8500000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.