Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mesityl oxide, 90+%, remainder 4-methyl-4-penten-2-one

Catalog No. aaa15111ak
Click to view available options
Quantity:
100 mL
250 mL
500 mL
1000 mL

CAS: 141-79-7 | C6H10O | 98.145 g/mol

Mesityl oxide is used as a solvent and in the production of methyl isobutyl ketone by hydrogenation, in the synthesis of 2,3,4,5-tetrahydro-1,3,3-trimethyldipyrrin, a crucial building block in the rational synthesis of chlorins and oxochlorins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Mesityl oxide is used as a solvent and in the production of methyl isobutyl ketone by hydrogenation, in the synthesis of 2,3,4,5-tetrahydro-1,3,3-trimethyldipyrrin, a crucial building block in the rational synthesis of chlorins and oxochlorins.

Solubility
Soluble in water(30g/L).

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-79-7
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.145
MDL Number MFCD00008900
InChI Key SHOJXDKTYKFBRD-UHFFFAOYSA-N
Synonym mesityl oxide, 4-methyl-3-penten-2-one, 3-penten-2-one, 4-methyl, isopropylideneacetone, isobutenyl methyl ketone, methyl isobutenyl ketone, mesityloxid, mesityloxyde, 3-isohexen-2-one, isopropylidene acetone
PubChem CID 8858
IUPAC Name 4-methylpent-3-en-2-one
SMILES CC(=CC(=O)C)C

Specifications

Melting Point °C to 59°C
Density 0.856
Boiling Point 129°C to 130°C
Flash Point 30°C (86°F)
Assay Percent Range ≥90%
Refractive Index 1.444
Quantity 250 mL
UN Number UN1229
Beilstein 1361550
Merck Index 14,5908
Solubility Information Soluble in water(30g/L).
Formula Weight 98.15
Percent Purity ≥90%
Physical Form Liquid
Assay remainder 4-methyl-4-penten-2-one
Chemical Name or Material Mesityl Oxide
Show More Show Less
Hazard Category H226-H302+H312+H332
Hazard Statement GHS H Statement
H226-H302-H312-H332
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P312-P330-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: MESITYL OXIDE
EINECSNumber 205-502-5
RTECSNumber SB4200000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.